Masterflex® L/S® Easy-Load® II Pump Head Tubing Retainer Kit for Precision Tubing
Catalog # MFLX77200-90
Supplier: Avantor Fluid Handling
About this item
Enhance the life of your pump head with the replacement retainer kits.
- Retainer kits provide the parts necessary to replace the spring-loaded tubing retainers in your Masterflex® Easy-Load® II pump heads
- Suitable for use with Easy-Load® II pump heads 77200-50, -60, and 77201-60
Replacement parts to keep your pump head operating even longer. Tubing retainers hold the tubing in place within the pump head. Pump head sold separately.
Delivery: Includes two small upper retainers, two small lower retainers, and two compression springs.
- Descripción:Tubing Retainer Kit for L/S Easy-Load II Pump Heads 77200-50, -60 and 77201-60,
- Environmentally Preferable:
- Garantía:1 year
- Referencia:MFLX77200-90
Frequently Bought Together