Masterflex® L/S® Easy-Load® II Pump Head Replacement Rotor Assembly for Precision Tubing, CRS
Catalog # MFLX77200-58
Supplier: Avantor Fluid Handling
About this item
Get the most our of your Masterflex® L/S® Easy-Load® II Pump Head with these replacement parts.
- Choose replacement parts suitable for your L/S® pump head
- Suitable for use with Easy-Load® II pump head 77200-50
Replace your pump head rotor assembly to extend the life of your pump head.
- Pump series:L/S
- Descripción:Replacement rotor assembly for L/S Easy-Load II pump heads 77200-50,
- Environmentally Preferable:
- Garantía:2 years
- Referencia:MFLX77200-58
- Rotor Material:Cold-rolled steel
Frequently Bought Together