Suction Line Pump Filter Strainers
Suction Line Strainer, PVC, 1/2" NPT(F), 50 Mesh (stainless steel), 0.5 NPT(F)
MFLX74604-23
: 3991550
Protect pump from sediments filtering into suction line.
- PVC (polyvinyl chloride) or Nylon base material of construction
- 20, 40, 80 mesh sizes available
- Easily removable for cleaning or replacing
These suction line pump filter strainers are ideal for keeping sediment and debris out of your pump for optimal performance. Choose from PVC (polyvinyl chloride) or nylon base material in a wide range of mesh sizes for any application.
- Description:Suction Line Strainer, PVC, 1/2" NPT(F), 50 Mesh (stainless steel)
- Inlet Connection:0.5 NPT(F)
- Cat. No.:MFLX74604-23
- Warranty:6 months
- Wetted materials:PVC and Stainless Steel
- Outlet connection:0.5 NPT(F)