ITT Jabsco Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
Catálogo # MFLX74021-04
Proveedor: BPH PUMP & EQUIPMENT INC
Acerca de este artículo
Create a pulse-free flow out of your positive displacement pump.
- Choose between PTFE or Polypropylene with Santoprene dampners
Dampeners work by pre-filling the chamber with compressed air or other gas, then as fluid pressure increases it compresses the air/gas in the chamber. When the air/gas pressure exceeds the fluid pressure the fluid is forced back out of the dampener in between pulse cycles, resulting in a near pulseless flow.
More Info: These pulse dampeners can be used with any positive displacement pump that needs flow with reduced pulsation.
- Descripción:Polypropylene Pulse Dampeners With Santoprene Diaphragm, 3/8" NPT
- Environmentally Preferable:
- Referencia:MFLX74021-04
- Wetted materials:Polypropylene, Santoprene®
- Conexión de entrada de aire:0.25 NPT(F)
- Inlet connection:0.375 NPT(F)
- Outlet connection:0.375 NPT(F)
- Garantía:1 year
Frequently Bought Together