$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
[EN]FLOWMETER 1-10 GPM WATER 1"NPT
MFLX61550-00
:
KEY INSTRUMENTS, LLC
[EN]FLOWMETER 1-10 GPM WATER 1"NPT
MFLX61550-00
:
KEY INSTRUMENTS, LLC
:
2540A5L57PI-CP
Frequently Bought Together
Mortar and Pestle Sets, Glass, Variety Glass
: VARIETY GLASS INC
Magnetic Stirring Bar Retriever
: VWR International
Accessories for Two-Speed Laboratory Blenders, 1 L, Waring
: WARING PRODUCTS DIVISION
BARRA MAGNETICA 50 X 20 MM ELIPTICA
: BRAND GMBH & CO. MX