Cadence Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Slip to 1/4" Barb
Catalog # MFLX41507-60
Supplier: CADENCE INC.
About this item
Excellent durability.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Manufactured in an ISO 14644-1 Level 8 (formerly Class 100,000) clean room
- Full material traceability on all items
Constructed of durable plated brass, these Luer Adapter Fittings are ideal for a wide range of applications.
- Tamaño de la púa/Ø int. del tubo (puerto 1):¼"
- Tipo de conexión:Male Luer to Barb
- Flow pattern:Straight
- Descripción:Luer Fitting Adapter, Nickel-Plated Brass, Male Luer Slip to 1/4" Barb
- Environmentally Preferable:
- Connection type (port 1):Male luer slip
- Connection type (port 2):1/4" Barb
- No. of ports:2
- Garantía:None
- Referencia:MFLX41507-60
- Fitting Material:Brass, nickel-plated
Frequently Bought Together