John Guest polypropylene threaded elbows
Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk, Push-to-Connect to Thread, Elbow, ¼", PP, ⅜"
Catalog # MFLX34007-64
Supplier: RELIANCE WORLDWIDE CORPORATION
Supplier #: PP481222W
John Guest polypropylene threaded elbows
Catalog # MFLX34007-64
Supplier: RELIANCE WORLDWIDE CORPORATION
Jump to:
Product Details & Documents
Quick, leakproof connection.
- Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications; and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.
Especificaciones
- Connection Type:Push-to-Connect to Thread
- Flow pattern:Elbow
- Thread size NPT(M) (port 2):¼"
- Description:Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk
- Fitting Material:PP
- Push-to-connect Tube O.D. (Port 1):⅜"
- Cat. No.:MFLX34007-64
- Warranty:20 years
- Min. temperature:1 °C (33 °F)
- Max. temperature:60 °C (140 °F)
- Sterilisation:Autoclave only
- Max. operating pressure:150 psi at 70 °F, 60 psi at 140 °F (for water)
- Seal material:EPDM
Accessories
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/8" NPT(M); 10/PkPush-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/8" NPT(M); 10/PkMFLX34007-53Push-to-Connect to ThreadStraightLog in Para disponibilidadEach (10UN) - $316.00
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/4" NPT(M); 10/PkPush-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/4" NPT(M); 10/PkMFLX34007-54Push-to-Connect to ThreadStraightLog in Para disponibilidadEach (10UN) - $316.00
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×1/4" NPT(M); 10/PkPush-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×1/4" NPT(M); 10/PkMFLX34007-55Push-to-Connect to ThreadStraightLog in Para disponibilidadEach (10UN) - $311.00
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×3/8" NPT(M); 10/PkPush-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×3/8" NPT(M); 10/PkMFLX34007-56Push-to-Connect to ThreadStraightLog in Para disponibilidadEach (10UN) - $381.00
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ¼" OD×¼" OD; 10/PkPush-To-Connect Fitting, Elbow Adapter, Polypropylene, ¼" OD×¼" OD; 10/PkMFLX34007-58Push-to-Connect UnionElbowLog in Para disponibilidadEach (10UN) - $734.00
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-to-connect reducer adapters, polypropylene, 3/8"×1/4", 10/packPush-to-connect reducer adapters, polypropylene, 3/8"×1/4", 10/packMFLX34007-57Push-to-Connect ReducerStraightLog in Para disponibilidad
Información del producto
- Catalog #Connection TypeFlow patternAvailabilityPrice
- Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ⅜" OD×⅜" OD; 10/PkPush-To-Connect Fitting, Elbow Adapter, Polypropylene, ⅜" OD×⅜" OD; 10/PkMFLX34007-60Push-to-Connect UnionElbowLog in Para disponibilidadEach (10UN) - $1,130.00