Orders LinkContactUsLinkComponent
John Guest polypropylene threaded elbows
John Guest polypropylene threaded elbows
Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk, Push-to-Connect to Thread, Elbow, ¼", PP, ⅜"
Catalog # MFLX34007-64
Supplier #:  PP481222W
John Guest polypropylene threaded elbows
Catalog # MFLX34007-64
John Guest polypropylene threaded elbows
John Guest polypropylene threaded elbows
Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk, Push-to-Connect to Thread, Elbow, ¼", PP, ⅜"
Price
$960.00
Log in Para disponibilidad
Each (10UN)
Product Details & Documents
Quick, leakproof connection.
  • Polypropylene and EPDM materials meet NSF 51, and NSF 61 requirements for food-grade applications; and is FDA compliant
Push-to-connect fittings offer secure connections and quick disconnection. Simply push tubing past the O-ring to the internal tube stop—steel teeth on the collet grip tubing for leakproof connections. To disconnect, press collet against fitting body to disengage steel teeth—tubing slides right out.
Fittings work with semi-rigid plastic (polypropylene, polyethylene, nylon) and soft metal (brass, copper, mild steel) tubing.
Adapters (sold separately) let you permanently modify basic fittings for customized installations.
Especificaciones
  • Connection Type:
    Push-to-Connect to Thread
  • Flow pattern:
    Elbow
  • Thread size NPT(M) (port 2):
    ¼"
  • Description:
    Threaded Elbow Adapters, 3/8" OD to 1/4" NPT(M), PP, 10/Pk
  • Fitting Material:
    PP
  • Push-to-connect Tube O.D. (Port 1):
    ⅜"
  • Cat. No.:
    MFLX34007-64
  • Warranty:
    20 years
  • Min. temperature:
    1 °C (33 °F)
  • Max. temperature:
    60 °C (140 °F)
  • Sterilisation:
    Autoclave only
  • Max. operating pressure:
    150 psi at 70 °F, 60 psi at 140 °F (for water)
  • Seal material:
    EPDM
Accessories
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/8" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/8" NPT(M); 10/Pk
    MFLX34007-53
    Push-to-Connect to Thread
    Straight
    Log in Para disponibilidad
    Each (10UN) - $316.00
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/4" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 1/4" OD×1/4" NPT(M); 10/Pk
    MFLX34007-54
    Push-to-Connect to Thread
    Straight
    Log in Para disponibilidad
    Each (10UN) - $316.00
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×1/4" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×1/4" NPT(M); 10/Pk
    MFLX34007-55
    Push-to-Connect to Thread
    Straight
    Log in Para disponibilidad
    Each (10UN) - $311.00
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×3/8" NPT(M); 10/Pk
    Push-To-Connect Fitting, Male Thread, Polypropylene, 3/8" OD×3/8" NPT(M); 10/Pk
    MFLX34007-56
    Push-to-Connect to Thread
    Straight
    Log in Para disponibilidad
    Each (10UN) - $381.00
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ¼" OD×¼" OD; 10/Pk
    Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ¼" OD×¼" OD; 10/Pk
    MFLX34007-58
    Push-to-Connect Union
    Elbow
    Log in Para disponibilidad
    Each (10UN) - $734.00
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-to-connect reducer adapters, polypropylene, 3/8"×1/4", 10/pack
    Push-to-connect reducer adapters, polypropylene, 3/8"×1/4", 10/pack
    MFLX34007-57
    Push-to-Connect Reducer
    Straight
    Log in Para disponibilidad
    Each (10UN) - $782.00
Información del producto
  • Catalog #
    Connection Type
    Flow pattern
    Availability
    Price
  • Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ⅜" OD×⅜" OD; 10/Pk
    Push-To-Connect Fitting, Elbow Adapter, Polypropylene, ⅜" OD×⅜" OD; 10/Pk
    MFLX34007-60
    Push-to-Connect Union
    Elbow
    Log in Para disponibilidad
    Each (10UN) - $1,130.00