Order Entry
Mexico
ContactUsLinkComponent
Hose Fittings, Band Clamps, for 1" OD Hose, 25/Pk
Hose Fittings, Band Clamps, for 1" OD Hose, 25/Pk
Catálogo # MFLX33350-12
Proveedor:  VWR International
undefined
Hose Fittings, Band Clamps, for 1" OD Hose, 25/Pk
Catálogo # MFLX33350-12
Proveedor:  VWR International

Acerca de este artículo

Quick and easy assembly with band clamp tool.

  • Constructed of durable stainless steel

Band Clamps are ideal for tightening hose over barbed fittings. Constructed of stainless steel. All band clamps require a band clamp tool to install.

  • Tubo ext.Ø:
    25.4 mm (1")
  • Descripción:
    Hose Fittings, Band Clamps, for 1" OD Hose, 25/Pk
  • Environmentally Preferable:
  • Garantía:
    1 year
  • Referencia:
    MFLX33350-12