Order Entry
Mexico
ContactUsLinkComponent
Hedland Water Flowmeter w/ Transmitter, 0.2-2.0 GPM, SS, 1/2" NPT(F)
Hedland Water Flowmeter w/ Transmitter, 0.2-2.0 GPM, SS, 1/2" NPT(F)
Catálogo # MFLX32509-86
Proveedor:  BADGER METER INC
undefined
Hedland Water Flowmeter w/ Transmitter, 0.2-2.0 GPM, SS, 1/2" NPT(F)
Catálogo # MFLX32509-86
Proveedor:  BADGER METER INC
Número de proveedor:  H605S-002-MR

Acerca de este artículo

Easy to read scale.

  • Direct reading
  • Install in any position
  • 360º Rotatable Guard/Scale
  • Easier ot Read Linear Scale
  • No Flow Straighteners or Special Piping Required
  • Insensitive to Shock and Vibration
  • Good Viscosity Stability
  • Temperature up to 240ºF
  • Accuracy ±2% Full Scale
  • Repeatability ±1%
  • Special Scales Available

Ideal for applications that are sensitive to shock and vibration.

  • Descripción:
    Water Flowmeter w/ Transmitter. 0.2-2.0 gal./min. SS. 1/2" NPT(F)
  • Environmentally Preferable:
  • Temperatura máximo de operación:
    115.5 °C (240 °F)
  • Garantía:
    1 year
  • Referencia:
    MFLX32509-86
Frequently Bought Together