ContactUsLinkComponent
Masterflex® Mass Flow Controller Power Supply; 110 VAC to 12 VDC
Masterflex® Mass Flow Controller Power Supply; 110 VAC to 12 VDC
Catálogo # MFLX32498-40
Masterflex® Mass Flow Controller Power Supply; 110 VAC to 12 VDC
Catálogo # MFLX32498-40
Número de proveedor:  PS-GFC-110NA-2
Acerca de este artículo
Make the most of your Masterflex flowmeters with these replacement parts and other accessories.
  • Optimize performance of your flowmeter with these accessories
These power supplies, adapters, and other accessories are suitable for use with your Masterflex flowmeter and flow sensors.
  • Descripción:
    Davis 110Vac/12VDC For Mass Flow Controller
  • Environmentally Preferable:
  • Modelo:
    32498-40
  • Garantía:
    1 year
  • Referencia:
    MFLX32498-40
Frequently Bought Together