Replacement PFA Compression Ferrule Nut and Assemblies
Compression Nut Assembly, PFA, ⅜", Compression Nut Assembly with Ferrule, Straight, PFA
MFLX31321-87
Supplier #: 1202-0002
Create a secure connection with compression fittings.
- Constructed of chemically inert PFA
Compression fittings are an excellent choice to connect rigid tubing systems and withstand higher pressures than barbed fittings. Assemble the fitting to the tubing by sliding the tubing through a sealing ferrule and using the nut to compress down to form a secure seal.
- Connection Type:Compression Nut Assembly with Ferrule
- Flow pattern:Straight
- Description:Compression Nut Assembly, PFA, ⅜"
- Fitting Material:PFA
- Cat. No.:MFLX31321-87
- Warranty:18 months
- Sterilisation:Autoclave or ethylene oxide (ETO)