Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded nipple, PFA, ¼" male NPT Fitting, Threaded Union, Straight, ¼", ¼", PFA
Supplier #: PN-4-P
- Connection Type:Threaded Union
- Flow pattern:Straight
- Thread size NPT(M) (port 1):¼"
- Thread size NPT(M) (port 2):¼"
- Description:Threaded nipple, PFA, ¼" male NPT Fitting
- Fitting Material:PFA
- Cat. No.:MFLX31321-00
- Warranty:18 months
- Sterilisation:Autoclave or ethylene oxide (ETO)