Orders LinkContactUsLinkComponent
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded nipple, PFA, ¼" male NPT Fitting, Threaded Union, Straight, ¼", ¼", PFA
  MFLX31321-00
CE certificate
Supplier #:  PN-4-P
Threaded Reducing Nipples, NPT(M)×NPT(M)
  MFLX31321-00
CE certificate
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded Reducing Nipples, NPT(M)×NPT(M)
Threaded nipple, PFA, ¼" male NPT Fitting, Threaded Union, Straight, ¼", ¼", PFA
$594.00
Each (1UN)
Create a secure connection with threaded fittings.
  • Constructed of chemically inert PFA
Threaded fittings and connections offer a more permanent installation. Use these nipples to reduce or enlarge connection sizes.
Documents
  • Connection Type:
    Threaded Union
  • Flow pattern:
    Straight
  • Thread size NPT(M) (port 1):
    ¼"
  • Thread size NPT(M) (port 2):
    ¼"
  • Description:
    Threaded nipple, PFA, ¼" male NPT Fitting
  • Fitting Material:
    PFA
  • Cat. No.:
    MFLX31321-00
  • Warranty:
    18 months
  • Sterilisation:
    Autoclave or ethylene oxide (ETO)