Masterflex® Fitting, Brass, Straight, Hosebarb to Thread Adapter, 3/8" ID x 3/8" NPT(M); 5/PK
Catalog # MFLX30904-21
Supplier: VWR International
About this item
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: ³/₈" int.Ø × ³/₈" NPT(M)
Available in a variety of materials and configurations to meet your need. Adapts from a NPT male thread to a hose barb.
- Tamaño de la púa/Ø int. del tubo (puerto 2):⅜"
- Tipo de conexión:Thread to Barb
- Flow pattern:Straight
- Thread size NPT(M) (port 1):⅜"
- Descripción:Fitting, Brass, Straight, Hosebarb to Thread Adapter, ⅜" ID × ⅜" NPT(M); 5/PK
- Environmentally Preferable:
- Modelo:30904-21
- No. of ports:2
- Garantía:1 year
- Referencia:MFLX30904-21
- Fitting Material:Brass
Frequently Bought Together