Masterflex® Fitting, Nylon, Tee, Hosebarb to Thread, 3/8" ID x 3/8" ID x 1/4" NTP(M); 10/PK
Catálogo # MFLX30624-07
Proveedor: VWR International
Acerca de este artículo
Quick and easy assembly.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
These tee adapters help you install your flexible tubing lines into more rigid, threaded lines. Choose the size and material to suit your application needs.
- Tamaño de la púa/Ø int. del tubo (puerto 2):⅜"
- Tipo de conexión:Thread to Barb
- Flow pattern:T
- Descripción:Fitting, Nylon, Tee, Hosebarb to Thread, 3/8" ID x 3/8" ID x 1/4" NTP(M); 10/PK
- Environmentally Preferable:
- Modelo:30624-07
- Orifice dimensions:0.25"
- Sterilisation:Ethylene oxide (ETO) only
- Referencia:MFLX30624-07
- Fitting Material:Nylon
Frequently Bought Together