Masterflex® Gold-colored IDentifier Hinge Clamp, 1/2-3/4" Clamp Size, 304 SS; 1/EA
Catálogo # MFLX30562-51
Proveedor: VWR International
Acerca de este artículo
Easy visual identification of critical joints for review in your process.
- Constructed of gold-colored 304 Stainless Steel
Easily distinguishable from standard stainless steel clamps from a distance, these gold-colored sanitary clamps are identical in size and function and can be used interchangeably with standard stainless steel clamps. Clamps feature a TiN (titanium nitride) coating. Clamps are available in a variety of sizes.
- Connection type:Sanitary
- Descripción:Gold-colored IDentifier Hinge Clamp, 1/2-3/4" Clamp Size, 304 SS; 1/EA
- Matriz:Stainless steel 304
- Environmentally Preferable:
- Sanitary clamp size (port 1):3/4"
- Garantía:1 year
- Referencia:MFLX30562-51
- Clamp Type:Sanitary
Frequently Bought Together