Masterflex® L/S® Peristaltic Pump Drive Replacement Brushes; 2/PK
MFLX07533-01
Quickly and easily replace components of your Masterflex system.
- Choose replacement brushes and caps based on your Masterflex L/S drive
- Brushes should be inspected every six months or 2000 hours—whichever comes first
Easy to install, these replacement drive parts will help extend the life of your drive.
- Descripción:L/S® Replacement Brushes, 2/PK
- Environmentally Preferable:
- Referencia:MFLX07533-01
Frequently Bought Together