ContactUsLinkComponent
Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Catálogo # MFLX07079-15
Proveedor:  ACTION PUMP COMPANY
undefined
Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
Catálogo # MFLX07079-15
Proveedor:  ACTION PUMP COMPANY
Número de proveedor:  258
Acerca de este artículo
Compatible with Siphon and Piston Hand-Operated Pumps.
  • Choose from a wide range of replacement parts and accessories
Model 07079-10 steel screw in adapter features a 1-3/4" NPT(F) connection and fits 5 gallon pails.
Model 07079-15 flex spout adapter is constructed of durable polyethylene and fits 5 gallon containers with 5-5/8" (6.7 cm) openings.
Model 07079-17 bung converter kit converts a Mauser course bung thread or standard course buttress to a 2" NPT connection.
  • Descripción:
    Flex Spout Adapter for 5-Gallon Containers with 2-5/8" Opening, polyethylene
  • Environmentally Preferable:
  • Garantía:
    1 year
  • Referencia:
    MFLX07079-15
Frequently Bought Together