Micropump Pump Head Adapter Kits
Gear Pump Head Adapter Kit, 56C face drive, standard magnets
Catalog # MFLX07003-99
Supplier: MICROPUMP INC
Número de proveedor #: 81518
Jump to:
Product Details & Documents
Easily adapt your pump head to your drive with these adapter kits.
- Choose adapter kit suitable for use with your Masterflex pump head
Use these adapter kits to attach Micropump® A-Mount pump heads to your own motor or Masterflex® L/S® pump drives.
Especificaciones
- Description:Gear Pump Head Adapter Kit, 56C face drive, standard magnets