Masterflex® Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
Catalog # MFLX06833-00
Supplier: VWR International
About this item
Won't rust, corrode, or distort.
- Constructed of durable Celcon plastic
Durable Celcon plastic clamp is autoclavable; won't rust, corrode, or distort. Accepts flexible tubing.
Mini clamp accept up to 1/4" OD tubing and provides on/off control. The standard clamp accepts up to 1/2" OD tubing and features twelve-position ratchet control to adjust flow from fully-open to complete-closure.
- Housing material:Polyoxymethylene (POM) Acetal
- Descripción:Ratchet Tubing Clamp for up to 1/2" Tubing, Acetal; 25/pack
- Environmentally Preferable:
- Pinch valve tube size:1/8" to 1/2" OD
- Garantía:3 Months
- Referencia:MFLX06833-00
Frequently Bought Together