Sanitary End Caps, Polypropylene
Sanitary End Cap, PP; 3" Sanitary Tri-Clamp, Sanitary Cap, Straight, 3", PP, 3"
Catalog # MFLX06320-83
Supplier: ST. GOBAIN - BEAVERTON
Número de proveedor #: TEC300
Jump to:
Product Details & Documents
Create a secure connection in your sanitary fitting system.
- Constructed of durable polypropylene (PP)
Close off your sanitary fluid line with these convenient end caps.
Especificaciones
- Connection Type:Sanitary Cap
- Flow pattern:Straight
- Sanitary clamp size (Port 1):3"
- Description:Sanitary End Cap, PP; 3" Sanitary Tri-Clamp
- Fitting Material:PP
- Sanitary Gasket Size:3"
- Cat. No.:MFLX06320-83
- Warranty:6 Months
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



