Burkle 5606-1000 Hand Operated Drum Pump with Spigot, PTFE/FEP, 95 cm inlet tube, 400 ml/stroke
Catálogo # MFLX06043-98
Proveedor: BURKLE INC
Acerca de este artículo
Safely transfer aggressive chemicals.
- PTFE and FEP wetted materials withstand harsh chemicals
- Stopcock contains hazardous vapors and prevents dripping
These PTFE barrel pumps are resistant to strong acids, bases, organics, and other chemicals (except elemental fluorine) and are equally suited to ultrapure liquid handling. Choose between pumps with a spout and a discharge hose, depending on your application. All models include a stopcock and require an adapter for use (order separately).
- Velocidad de flujo:2.5 strokes/L (9.5 strokes/gal.)
- Descripción:Hand Operated Drum Pump with Spigot, PTFE/FEP, 95 cm inlet tube, 400 ml/stroke
- Environmentally Preferable:
- Wetted materials:PTFE/FEP
- Outlet connection:Spigot
- Tube ext.Ø, inlet:3.2 cm (1.25")
- Tube length, inlet:95 cm (37.4")
- Garantía:15 Months
- Referencia:MFLX06043-98