ContactUsLinkComponent
Masterflex® Tripod Base for Multi-Tube Frames; 1/Each
Masterflex® Tripod Base for Multi-Tube Frames; 1/Each
Catálogo # MFLX03226-50
undefined
Masterflex® Tripod Base for Multi-Tube Frames; 1/Each
Catálogo # MFLX03226-50
Número de proveedor:  TP2
Acerca de este artículo
Easily mount flowmeters on tripod for easy viewing.
  • Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
  • Descripción:
    Tripod Base for Multi-Tube Frames
  • Environmentally Preferable:
  • Modelo:
    03226-50
  • Ancho:
    1755"
  • Garantía:
    1 year
  • Referencia:
    MFLX03226-50
  • Housing Material:
    Acrylic
Frequently Bought Together