Masterflex® Tripod Base for Multi-Tube Frames; 1/Each
Catálogo # MFLX03226-50
Proveedor: AALBORG INSTRUMENTS & CONTROL
Acerca de este artículo
Easily mount flowmeters on tripod for easy viewing.
- Constructed of durable acrylic
These tripod bases are ideal for benchtop applications. A tripod base is required to use the multitube frame with the common inlet flow pattern; all other multitube frames are designed for panel and tripod mounting. Tripod bases are made of acrylic and include a spirit leveler.
- Descripción:Tripod Base for Multi-Tube Frames
- Environmentally Preferable:
- Modelo:03226-50
- Ancho:1755"
- Garantía:1 year
- Referencia:MFLX03226-50
- Housing Material:Acrylic
Frequently Bought Together