Masterflex® Nylon Clamp for Sanitary Fittings, 1/2-3/4" Clamp Size; 10/PK
Catálogo # MFLX00424-TN
Proveedor: VWR International
Acerca de este artículo
Collared clamp helps prevent unintentional opening.
- All fittings are compatible with Tri-Clamp® fittings
- Excellent chemical resistance; low leachable characteristics
- Connection type:Sanitary
- Descripción:Nylon Clamp for Sanitary Fittings, 1/2-3/4" Clamp Size; 10/PK
- Matriz:Nylon
- Environmentally Preferable:
- Sanitary clamp size (port 1):1/2"
- Sterilisation:Autoclave, Gamma
- Garantía:1 year
- Referencia:MFLX00424-TN
- Clamp Type:Sanitary
Frequently Bought Together