$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
[EN]CASTOR CASSETTE BLOCK
MFLX00369-PB
:
Avantor Fluid Handling
[EN]CASTOR CASSETTE BLOCK
MFLX00369-PB
:
Avantor Fluid Handling
:
IS2115
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH