Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
REGLA RIGIDA AC.INOX MITYO182-111
85018-812
:
GRAINGER,S.A. DE C.V. MX
REGLA RIGIDA AC.INOX MITYO182-111
85018-812
:
GRAINGER,S.A. DE C.V. MX
:
4CGU9
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH