Skip to Header
Skip to Main Content
Skip to Footer
Avanzado
Avanzado
SDS
Certificados
Estructura química
Datos del fabricante
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Mexico
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
DESINFECTANTE ESTERILIZANTE CS4
Catalog # 85017-812
Supplier:
CYTNIS S.A DE C.V MX
Expand image
DESINFECTANTE ESTERILIZANTE CS4
Catalog # 85017-812
Supplier:
CYTNIS S.A DE C.V MX
Supplier Number:
6525-08EC
Frequently Bought Together
VWR® Light-Duty Tissue Wipers
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, Polypropylene
Supplier: VWR International
VWR® Nitrile Examination Gloves, Blue
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
Supplier: BDH