Orders LinkContactUsLinkComponent
Long-Handled Water Dippers
Long-Handled Water Dippers
Sample dipper with handle, 6'
Catalog # 470323-316
Supplier:   BEL-ART PRODUCTS SE
Número de proveedor #:  F36785-0016
Long-Handled Water Dippers
Catalog # 470323-316
Long-Handled Water Dippers
Long-Handled Water Dippers
Sample dipper with handle, 6'
Price
$2,663.04
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Ideal for sampling ponds and lakes.
  • Two pouring spouts on cup allow left or right pouring
  • High-density polyethylene plastic handles are strong honeycomb tubes without inserts
  • Handles are ribbed for a sure grip
  • Handle end has a hook for hanging storage
Collect water samples while remaining on the shore; the long handles of the water dippers let you reach three or six feet from dry land. Lightweight and easy to use, each dipper is made of durable plastic to withstand years of use. The molded 16 oz. cup screws onto the handle and has two pouring spouts for added versatility. Available in two convenient lengths.
Especificaciones
  • Description:
    Sample dipper with handle
  • Handle Length:
    6'
  • Cat. No.:
    470323-316