Order Entry
Mexico
Orders LinkContactUsLinkComponent
Ward's® Caliche

Ward's® Caliche

Caliche, bulk pack, 1 kg, Size will vary
Catalog # 470323-126
Supplier:   Avantor
Ward's® Caliche
Catalog # 470323-126
Supplier:  Avantor
Ward's® Caliche
Ward's® Caliche

Caliche, bulk pack, 1 kg, Size will vary

Price
$482.65
Log in for availability
Paquete de 1
Product Details & Documents

Hardened natural cement of calcium carbonate that binds other material such as gravel, sand, clay, and silt.

  • Collected in Arizona

  • Multiple sizes and pack options to fit your specific needs
  • Cut by Ward's geologists to guarantee top market quality
  • Matching thin section available

Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.

Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.

All specimen dimensions are considered an average size range.

Specifications
  • Description:
    Caliche, bulk pack, 1 kg
  • Specimen Dimensions:
    Size will vary