Ward's® Caliche
Caliche, student specimens, 1×1 - 1×1"
Catalog # 470323-116
Supplier: Avantor
Jump to:
Product Details & Documents
Hardened natural cement of calcium carbonate that binds other material such as gravel, sand, clay, and silt.
Collected in Arizona
- Multiple sizes and pack options to fit your specific needs
- Cut by Ward's geologists to guarantee top market quality
- Matching thin section available
Student specimens - Perfect for study and identification, these budget-friendly samples come sealed in a plastic bag. Storage box (5×5×4.5"), sold separately.
Hand specimens - Ideal for lectures, demonstrations, and display. Each comes in a labeled plastic tray with a matching labeled storage box.
All specimen dimensions are considered an average size range.
Especificaciones
- Description:Caliche, student specimens
- Specimen Dimensions:1×1 - 1×1"