Orders LinkContactUsLinkComponent
HandyStep® touch Motorized Repeating Pipette, BrandTech® Scientific
HandyStep® touch Motorized Repeating Pipette, BrandTech® Scientific
HandyStep® touch S with AC adapter
Catalog # 76338-256
Supplier #:  705210
HandyStep® touch Motorized Repeating Pipette, BrandTech® Scientific
Catalog # 76338-256
Supplier:  
HandyStep® touch Motorized Repeating Pipette, BrandTech® Scientific
HandyStep® touch Motorized Repeating Pipette, BrandTech® Scientific
HandyStep® touch S with AC adapter
Discontinued
Product Details & Documents
The HandyStep® touch brings intuitive touchscreen control to pipetting. A large color display provides all pertinent information.
  • Uses all standard dispenser tips
  • Automatically recognizes tip size of BRAND® PD-Tip™ II dispenser tips
  • Performs dispensing, auto-dispensing and single volume pipetting functions
  • Auto-dispensing 'learn' function replicates user rhythm to reduce repetitive motion by up to 97% and to make dispensing intervals easy and intuitive
  • Motorized tip ejection
  • Multi-lingual interface
The BRAND® HandyStep® touch has a large color touchscreen display that provides smart-phone like performance in a stepper pipette. All important features number of steps, aspiration and dispensing speed and more are readily visible and easily adjusted, even with gloves. Fits all standard type repeating tips. Automatic tip recognition of BRAND® PD-Tip™ II dispenser tips maximizes ease of use and minimizes error.
Two models cover all functions. Dispensing can be performed either manually, or in an automatic mode, which dispenses at defined intervals to reduce repetitive motion. Up to ten favorite settings can be saved. The HandyStep® touch S features added capability. It performs multi-aspiration - to allow filling with set volumes of multiple liquids for a single dispense, or sequential-dispensing, which permits the dispensing of up to ten different set volumes. Both instruments feature a high-capacity lithium-ion battery and are supplied with a universal battery charger for international use. A shelf/rack mount allows for easy storage of the HandyStep® touch, also mounts to a Transferpette® S pipette carousel.
 : Accessories include an inductive charging stand, and a support stand - both of which allow for a PD-Tip™ to be mounted for use while in the stand.
 : Additional shelf/rack mounts, replacement batteries and chargers are also available.
Documents
Specifications
  • Description:
    HandyStep® touch S with AC adapter
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Inductive charging stand for HandyStep® touch, without AC adapter
    Inductive charging stand for HandyStep® touch, without AC adapter
    76338-258
    Log in for availability
    Each (1UN) - $13,480.25