$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Mexico
ContactUsLinkComponent
AMPLITUD LAMBDA NW CELULAR/POLY 12 X 12
85015-576
:
CONTEC, INC. MX
AMPLITUD LAMBDA NW CELULAR/POLY 12 X 12
85015-576
:
CONTEC, INC. MX
:
AMLA0012
Frequently Bought Together
VWR® Disposable Serological Pipettes
: VWR International
VWR® Universal Pipette Tips
: VWR International
VWR® Disposable Petri Dishes
: VWR International
VWR® Nitrile Examination Gloves, Blue
: VWR International
pH Reference Standard Buffers, VWR Chemicals BDH®
: BDH