Orders LinkContactUsLinkComponent
Fixed Blade Scalpels
Fixed Blade Scalpels
Stainless steel, Narrow Blade Scalpel
Catalog # 470315-248
Supplier #:  25
Fixed Blade Scalpels
Catalog # 470315-248
Fixed Blade Scalpels
Fixed Blade Scalpels
Stainless steel, Narrow Blade Scalpel
Price
$53.21
Log in for availability
Each (1UN) Caja de 50 (1UN)
Product Details & Documents
Secure blade attachment.
  • Stainless steel construction
  • Fixed blade for additional strength
Fixed blade scalpels typically are stronger and less likely break than disposable blades. Often used for heavy cutting jobs, fixed blades are also less likely to twist while cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    Narrow Blade Scalpel
  • Material:
    Stainless steel