Hemostatic Forceps
Hemostat, Stainless steel, 127 mm (5"), Curved
Catalog # 470315-204
Supplier: DR INSTRUMENTS INC. SE
Número de proveedor #: 45CD
Jump to:
Product Details & Documents
Precise and secure holding.
- Stainless steel construction
- Available in curved or straight points
- One handed operation
For dissection use, hemostats are usually used to hold tissue and pull it out of the way. The locking mechanism in the handle also makes them useful as a clamp to either close off small tubing, or hold items together.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Hemostat
- Length:127 mm (5")
- Material:Stainless steel
- Curvature:Curved