Orders LinkContactUsLinkComponent
T-Pins
T-Pins
1.5" T-Pins, Nickel Plated, 38 mm (1.5")
Catalog # 470315-202
Número de proveedor #:  42-6/PK
T-Pins
Catalog # 470315-202
T-Pins
1.5" T-Pins, Nickel Plated, 38 mm (1.5")
Price
$45.69
Log in Para disponibilidad
Paquete de 6 Caja de 50 (6UN)
Product Details & Documents
Generally used to hold tissue to the dissection pan, T-pins are also good for marking structures.
  • Two sizes available
  • Nickel plated
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
  • Description:
    1.5" T-Pins
  • Length:
    38 mm (1.5")
  • Material:
    Nickel Plated