Orders LinkContactUsLinkComponent
T-Pins
T-Pins
2" T Pins, Nickel Plated, 50 mm (2")
Catalog # 470315-200
Supplier #:  42-100/PK
T-Pins
Catalog # 470315-200
T-Pins
2" T Pins, Nickel Plated, 50 mm (2")
Price
$163.35
Log in for availability
Paquete de 100 Caja de 10 (100UN)
Product Details & Documents
Generally used to hold tissue to the dissection pan, T-pins are also good for marking structures.
  • Two sizes available
  • Nickel plated
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Specifications
  • Description:
    2" T Pins
  • Length:
    50 mm (2")
  • Material:
    Nickel Plated