Microdissection Forceps
Curved Forceps, Stainless steel, 115 mm (4.5"), Curved
Catalog # 470315-184
Supplier: DR INSTRUMENTS INC. SE
Número de proveedor #: 18C
Jump to:
Product Details & Documents
For extremely fine work.
- Highly precise forceps
- High quality stainless steel construction
High quality materials and excellent craftsmanship combine in these microdissection forceps. Available as a straight fine point, a curved, or a self locking these forceps will exceed your expectations.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
Especificaciones
- Description:Curved Forceps
- Length:115 mm (4.5")
- Material:Stainless steel
- Curvature:Curved