Orders LinkContactUsLinkComponent
Fixed Blade Scalpels
Stainless steel, Microdissection Scalpel
  470315-172
 :  52
Fixed Blade Scalpels
  470315-172
Fixed Blade Scalpels
Fixed Blade Scalpels
Stainless steel, Microdissection Scalpel
$66.66
Each (1UN) Caja de 25 (1UN)
Secure blade attachment.
  • Stainless steel construction
  • Fixed blade for additional strength
Fixed blade scalpels typically are stronger and less likely break than disposable blades. Often used for heavy cutting jobs, fixed blades are also less likely to twist while cutting.
Caution: These instruments are intended for training and educational use only. These are not intended for human or animal procedures.
  • Description:
    Microdissection Scalpel
  • Material:
    Stainless steel