Order Entry
Mexico
Orders LinkContactUsLinkComponent
Polystyrene Petri Dishes

Polystyrene Petri Dishes

Petri Dish, 90 mm, 15 mm, 90×15 mm, Sterile
Catalog # 470313-486
Número de proveedor #:  1500-10
Polystyrene Petri Dishes
Catalog # 470313-486
Polystyrene Petri Dishes
Polystyrene Petri Dishes

Petri Dish, 90 mm, 15 mm, 90×15 mm, Sterile

Price
$202.09
Log in Para disponibilidad
Paquete de 20 Case de 10 (20UN)
Product Details & Documents

These sterile, two-piece polystyrene Petri dishes are sold in sleeves of 20.

Disposable polystyrene.

Especificaciones
  • Ø:
    90 mm
  • Sterility:
    Sterile
  • Description:
    Petri Dish
  • Height:
    15 mm
  • Diameter ×H:
    90×15 mm