Orders LinkContactUsLinkComponent
Ward's LiveSlides®
LiveSlides® Protist Study Set
  470313-232
 :   Avantor
Ward's LiveSlides®
  470313-232
 :  Avantor
Ward's LiveSlides®
Ward's LiveSlides®
LiveSlides® Protist Study Set
$745.17
Paquete de 5
Never before has it been so easy to view microorganisms, including bacteria, fungi, and protists.
  • Available as bulk pack of 50 slides only, or pack of 5 slides with transfer pipettes
  • Uniform depth of viewing well perfect for maintaining a shallow depth of field
  • Partitions help keep specimens in the viewing area
  • Polycarbonate material is safe for indoor or outdoor use
  • Can be used with traditional or digital microscopes
Each LiveSlide® consists of a base slide, and a set of coverslips of different thicknesses. One is very thin (0.13 mm), perfect for high magnification, and the second is a bit thicker (0.25 mm) for all applications. Each coverslip has a thin plastic film on each side for protection, one green and one clear. Remove both films by either peeling the film off or using a small piece of tape on the corner and lifting. Discard the films.
  • Description:
    LiveSlides® Protist Study Set