Serological Pipettes, Class A
25 ml, ±0.030 ml, Blue
Catalog # 470312-904
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0573E
Jump to:
Product Details & Documents
Perfect pipette for everyday laboratory use.
- Reusable
- Color-coded for easy sorting
- Accuracy as per Class A and DIN ISO 835 standard
These class A serological pipettes are calibrated for delivery to jet. The last drop is expelled by blowing. Blue graduations and color coding make these pipettes very easy to use.
Especificaciones
- Capacity:25 ml
- Color Code:Blue
- Tolerance:±0.030 ml