Orders LinkContactUsLinkComponent
Serological Pipettes, Class A
Serological Pipettes, Class A
25 ml, ±0.030 ml, Blue
Catalog # 470312-904
Número de proveedor #:  CH0573E
Serological Pipettes, Class A
Catalog # 470312-904
Serological Pipettes, Class A
Serological Pipettes, Class A
25 ml, ±0.030 ml, Blue
Price
$338.60
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Perfect pipette for everyday laboratory use.
  • Reusable
  • Color-coded for easy sorting
  • Accuracy as per Class A and DIN ISO 835 standard
These class A serological pipettes are calibrated for delivery to jet. The last drop is expelled by blowing. Blue graduations and color coding make these pipettes very easy to use.
Especificaciones
  • Capacity:
    25 ml
  • Color Code:
    Blue
  • Tolerance:
    ±0.030 ml