Laboratory Aprons
Regular, 42×27"
Catalog # 470311-692
Supplier: UNITED SCIENTIFIC SUPPLIES
Supplier #: LBAP02
Jump to:
Product Details & Documents
Protective laboratory apron for multipurpose use.
- Flexible
- Chemical resistant
- Apron includes cloth ties at the neck and waist
These aprons are made of a PVC-coated fabric which stays flexible. There is reinforcement at the points of strain.
Specifications
- Garment fit:Regular
- L×W:42×27"
- Cat. No.:470311-692
- Garment application:Chemical protection|General Purpose|Laboratory
- Gender:Unisex
- Fabric:PVC-coated fabric