Ward's Lab Wipe Holders
Large Lab Wipe Holder, 320×127×97 mm
470311-040
: HS234522W
Holds lab wipes such as Kimwipes® and other major brands.
- Mounts on a shelf or wall
- Mounting materials included
Clear acrylic holders fit boxes of small or large lab wipes. The smaller version attaches with double-stick tape and the larger version with screws.
- Description:Large Lab Wipe Holder
- Dimensions:320×127×97 mm