Orders LinkContactUsLinkComponent
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
250 ml, 2.0 ml
Catalog # 470310-998
Supplier #:  CH0346E
Single Scale Graduated Cylinders
Catalog # 470310-998
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
250 ml, 2.0 ml
Price
$248.85
Log in for availability
Each (1UN)
Product Details & Documents
Single Scale Graduated Cylinders with plastic base and protection collar.
  • Class B
  • Borosilicate glass gradauted cylinder
  • Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
Specifications
  • Graduation Interval:
    2.0 ml
  • Capacity:
    250 ml