Single Scale Graduated Cylinders
100 ml, 1.0 ml
Catalog # 470310-994
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0346C
Jump to:
Product Details & Documents
Single Scale Graduated Cylinders with plastic base and protection collar.
- Class B
- Borosilicate glass gradauted cylinder
- Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
Especificaciones
- Graduation Interval:1.0 ml
- Capacity:100 ml