Orders LinkContactUsLinkComponent
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
100 ml, 1.0 ml
Catalog # 470310-994
Número de proveedor #:  CH0346C
Single Scale Graduated Cylinders
Catalog # 470310-994
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
100 ml, 1.0 ml
Price
$146.71
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Single Scale Graduated Cylinders with plastic base and protection collar.
  • Class B
  • Borosilicate glass gradauted cylinder
  • Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
Especificaciones
  • Graduation Interval:
    1.0 ml
  • Capacity:
    100 ml