Single Scale Graduated Cylinders
25 ml, 0.5 ml
470310-992
Supplier #: CH0346B
Single Scale Graduated Cylinders with plastic base and protection collar.
- Class B
- Borosilicate glass gradauted cylinder
- Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
- Graduation Interval:0.5 ml
- Capacity:25 ml