Orders LinkContactUsLinkComponent
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
25 ml, 0.5 ml
  470310-992
Supplier #:  CH0346B
Single Scale Graduated Cylinders
  470310-992
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
25 ml, 0.5 ml
$122.56
Each (1UN)
Single Scale Graduated Cylinders with plastic base and protection collar.
  • Class B
  • Borosilicate glass gradauted cylinder
  • Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
  • Graduation Interval:
    0.5 ml
  • Capacity:
    25 ml