Orders LinkContactUsLinkComponent
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
10 ml, 0.2 ml
Catalog # 470310-990
Número de proveedor #:  CH0346A
Single Scale Graduated Cylinders
Catalog # 470310-990
Single Scale Graduated Cylinders
Single Scale Graduated Cylinders
10 ml, 0.2 ml
Price
$116.09
Log in Para disponibilidad
Each (1UN)
Product Details & Documents
Single Scale Graduated Cylinders with plastic base and protection collar.
  • Class B
  • Borosilicate glass gradauted cylinder
  • Detachable plastic hexagonal base
This graduate cylinder is perfect for most laboratory and classroom uses, and provides extra breakage protection with plastic hexagonal base and collar.
Especificaciones
  • Graduation Interval:
    0.2 ml
  • Capacity:
    10 ml