Orders LinkContactUsLinkComponent
TPX Beakers
50, 100, 250, 600, 1000 ml (Set)
  470310-782
 :  CH0138PRSET
TPX Beakers
  470310-782
TPX Beakers
TPX Beakers
50, 100, 250, 600, 1000 ml (Set)
$1,941.81
Each (1UN)
These TPX (PMP) beakers are idea for general laboraotry use.
  • Chemically resistant
  • Autoclavable
  • Long lasting and high clarity beakers
Able to withstand temperatures up to 180 °C.
  • Capacity:
    50, 100, 250, 600, 1000 ml (Set)