Melting Point Capillary Tubes
1 Open End
Catalog # 470310-684
Supplier: EISCO SCIENTIFIC LLC
Número de proveedor #: CH0882B
Jump to:
Product Details & Documents
Borosilicate glass capillary melting tubes are 4" in length and have a 0.05" outer diameter capillary tubes that are particularly useful for determining the melting point of various substances.
The 100 capillary tubes come in a cylindrical container that has a hole in the lid for easily obtaining one tube at a time.
Especificaciones
- Description:1 Open End