Orders LinkContactUsLinkComponent
Neoprene Stoppers
Neoprene Stoppers
14
Catalog # 470310-680
Supplier #:  CH0321X
Neoprene Stoppers
Catalog # 470310-680
Neoprene Stoppers
Neoprene Stoppers
14
Price
$6,218.82
Log in Para disponibilidad
Paquete de 10
Product Details & Documents
Versatile neoprene stoppers perfect for every laboratory.
  • Solid, one hole or two hole stoppers are available
  • Manufacturered according to sizes recommended by ASTM standards
Manufactured from long lasting neoprene rubbers. Suitable for use with petroleum products, mineral oils, fats as well as most inorganic acids and bases. Neoprene rubber is oil resistant and suitable for temperatures up to 120 °C (250 °F).
Packaging: Packs of 10.
Especificaciones
  • Size:
    14