Melting Point Capillary Tubes
2 Open Ends
Catalog # 470310-650
Supplier: EISCO SCIENTIFIC LLC
Supplier #: CH0882A
Jump to:
Product Details & Documents
Borosilicate glass capillary melting tubes are 4" in length and have a 0.05" outer diameter capillary tubes that are particularly useful for determining the melting point of various substances.
The 100 capillary tubes come in a cylindrical container that has a hole in the lid for easily obtaining one tube at a time.
Specifications
- Description:2 Open Ends