Orders LinkContactUsLinkComponent
Melting Point Capillary Tubes
Melting Point Capillary Tubes
2 Open Ends
Catalog # 470310-650
Supplier #:  CH0882A
Melting Point Capillary Tubes
Catalog # 470310-650
Melting Point Capillary Tubes
Melting Point Capillary Tubes
2 Open Ends
Price
$141.90
Log in for availability
Paquete de 100
Product Details & Documents
Borosilicate glass capillary melting tubes are 4" in length and have a 0.05" outer diameter capillary tubes that are particularly useful for determining the melting point of various substances.
The 100 capillary tubes come in a cylindrical container that has a hole in the lid for easily obtaining one tube at a time.
Specifications
  • Description:
    2 Open Ends